Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr5a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF327373.1 | - | 2 - 3 | 218 | 80 | |
BC124756.1 | - | 2 - 3 | 311 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |