Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arrdc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080029.1 | NP_001073498.1 | 3 - 4 | 688 | 126 | |
GenBank mRNA | BC117667.1 | - | 3 - 4 | 688 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |