Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mxra8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080582.1 | NP_001074051.1 | 3 - 4 | 513 | 126 | |
XM_009306033.3 | XP_009304308.1 | 4 - 5 | 616 | 126 | ||
XM_021479639.1 | XP_021335314.1 | 4 - 5 | 617 | 126 | ||
GenBank mRNA | BC129223.1 | - | 3 - 4 | 513 | 126 | |
CU638765.1 | - | 3 - 4 | 513 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |