Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rorca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082819.1 | NP_001076288.1 | 6 - 7 | 1053 | 81 | |
GenBank mRNA | DQ017627.1 | - | 6 - 7 | 1053 | 81 | |
EF107094.1 | - | 6 - 7 | 1053 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |