Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: col11a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083844.1 | NP_001077313.1 | 16 - 17 | 1771 | 57 | |
XM_005162813.1 | XP_005162870.1 | 17 - 18 | 2098 | 57 | ||
XM_005162814.1 | XP_005162871.1 | 17 - 18 | 2095 | 57 | ||
XM_005162815.1 | XP_005162872.1 | 16 - 17 | 2011 | 57 | ||
XM_005162818.1 | XP_005162875.1 | 16 - 17 | 1801 | 57 | ||
XM_021470209.1 | XP_021325884.1 | 15 - 16 | 1714 | 57 | ||
XM_021470210.1 | XP_021325885.1 | 10 - 11 | 878 | 57 | ||
GenBank mRNA | BC133083.1 | - | 16 - 17 | 1771 | 57 | |
GQ485664.1 | - | 17 - 18 | 1992 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |