Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: col11a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083844.1 | NP_001077313.1 | 23 - 24 | 2127 | 90 | |
XM_005162813.1 | XP_005162870.1 | 24 - 25 | 2454 | 90 | ||
XM_005162814.1 | XP_005162871.1 | 24 - 25 | 2451 | 90 | ||
XM_005162815.1 | XP_005162872.1 | 23 - 24 | 2367 | 90 | ||
XM_005162818.1 | XP_005162875.1 | 23 - 24 | 2157 | 90 | ||
XM_021470209.1 | XP_021325884.1 | 22 - 23 | 2070 | 90 | ||
XM_021470210.1 | XP_021325885.1 | 17 - 18 | 1234 | 90 | ||
GenBank mRNA | BC133083.1 | - | 23 - 24 | 2127 | 90 | |
GQ485664.1 | - | 24 - 25 | 2348 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |