Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pfkmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089417.2 | NP_001082886.2 | 10 - 11 | 1133 | 63 | |
NM_001365336.1 | NP_001352265.1 | 13 - 14 | 1647 | 63 | ||
GenBank mRNA | BC139532.1 | - | 10 - 11 | 1120 | 63 | |
EB884905.1 | - | 3 - 4 | 249 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |