Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pfkmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089417.2 | NP_001082886.2 | 11 - 12 | 1206 | 92 | |
NM_001365336.1 | NP_001352265.1 | 14 - 15 | 1720 | 92 | ||
GenBank mRNA | BC139532.1 | - | 11 - 12 | 1193 | 92 | |
EB884905.1 | - | 4 - 5 | 322 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |