Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zp2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089502.1 | NP_001082971.1 | 4 - 5 | 838 | 76 | |
GenBank mRNA | BC134168.1 | - | 4 - 5 | 838 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |