Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zp2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089502.1 | NP_001082971.1 | 5 - 6 | 1036 | 88 | |
GenBank mRNA | BC134168.1 | - | 5 - 6 | 1036 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |