Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ankrd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102388.1 | NP_001095858.1 | 4 - 5 | 404 | 77 | |
GenBank mRNA | BC150367.1 | - | 4 - 5 | 404 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |