Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sema3aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131060.1 | NP_571135.1 | 14 - 15 | 2013 | 80 | |
GenBank mRNA | AF086761.1 | - | 14 - 15 | 2013 | 80 | |
BC162381.1 | - | 14 - 15 | 1726 | 80 | ||
BC162398.1 | - | 14 - 15 | 1726 | 80 | ||
CU458855.1 | - | 14 - 15 | 1656 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |