Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sema3aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131060.1 | NP_571135.1 | 2 - 3 | 631 | 93 | |
GenBank mRNA | AF086761.1 | - | 2 - 3 | 631 | 93 | |
BC162381.1 | - | 2 - 3 | 93 | |||
BC162398.1 | - | 2 - 3 | 344 | 93 | ||
CU458855.1 | - | 2 - 3 | 274 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |