Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pou6f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131113.1 | NP_571188.1 | 2 - 3 | 339 | 71 | |
GenBank mRNA | BC162134.1 | - | 2 - 3 | 181 | 71 | |
BC162146.1 | - | 2 - 3 | 181 | 71 | ||
X68432.1 | - | 339 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |