Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnmt3bb.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131386.1 | NP_571461.1 | 14 - 15 | 3265 | 103 | |
GenBank mRNA | AB196914.1 | - | 14 - 15 | 3265 | 103 | |
AF135438.1 | - | 10 - 11 | 1248 | 103 | ||
BC162467.1 | - | 14 - 15 | 3154 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |