Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnmt3bb.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131386.1 | NP_571461.1 | 21 - 22 | 4108 | 75 | |
GenBank mRNA | AB196914.1 | - | 21 - 22 | 4108 | 75 | |
AF135438.1 | - | 17 - 18 | 2091 | 75 | ||
BC162467.1 | - | 21 - 22 | 3997 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |