Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ebf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131418.2 | NP_571493.2 | 11 - 12 | 1575 | 60 | |
XM_005165618.4 | XP_005165675.1 | 11 - 12 | 1627 | 60 | ||
GenBank mRNA | AF072657.1 | - | 11 - 12 | 1535 | 60 | |
BC162504.1 | - | 11 - 12 | 1384 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |