Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131573.2 | NP_571648.1 | 2 - 3 | 202 | 80 | |
XM_017357715.2 | XP_017213204.1 | 2 - 3 | 241 | 80 | ||
GenBank mRNA | AF231128.1 | - | 2 - 3 | 80 | ||
BC091443.1 | - | 2 - 3 | 198 | 80 | ||
BC115168.1 | - | 2 - 3 | 202 | 80 |
Gene Symbol: LOC101885164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017357859.2 | XP_017213348.1 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |