Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arntl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131577.1 | NP_571652.1 | 18 - 19 | 1860 | 87 | |
XM_005163022.4 | XP_005163079.1 | 18 - 19 | 1927 | 87 | ||
GenBank mRNA | AF144689.1 | - | 18 - 19 | 1860 | 87 | |
AF144690.1 | - | 18 - 19 | 1877 | 87 | ||
BC044135.1 | - | 18 - 19 | 1859 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |