Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmb13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131677.2 | NP_571752.2 | 5 - 6 | 558 | 152 | |
GenBank mRNA | AF155577.1 | - | 5 - 6 | 510 | 152 | |
BC092899.1 | - | 5 - 6 | 152 | |||
BC165548.1 | - | 5 - 6 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |