Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cryaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152950.2 | NP_694482.1 | 1 - 2 | 292 | 127 | |
GenBank mRNA | AY035778.1 | - | 1 - 2 | 127 | ||
BC083177.1 | - | 1 - 2 | 292 | 127 | ||
BC164886.1 | - | 1 - 2 | 221 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |