Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dachb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152956.2 | NP_694488.2 | 5 - 6 | 1111 | 109 | |
XM_021477542.1 | XP_021333217.1 | 4 - 5 | 524 | 109 | ||
GenBank mRNA | AF427109.1 | - | 5 - 6 | 1085 | 109 | |
BC163528.1 | - | 5 - 6 | 1078 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |