Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: igf1ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152968.1 | NP_694500.1 | 2 - 3 | 660 | 62 | |
XM_017351896.2 | XP_017207385.1 | 2 - 3 | 1592 | 62 | ||
GenBank mRNA | AF400275.1 | - | 2 - 3 | 660 | 62 | |
BC163723.1 | - | 2 - 3 | 724 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |