Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnaia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198805.2 | NP_942100.1 | 3 - 4 | 663 | 69 | |
GenBank mRNA | BC049537.1 | - | 3 - 4 | 626 | 69 | |
EH537458.1 | - | 3 - 4 | 254 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |