Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jade1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199805.1 | NP_956099.1 | 3 - 4 | 231 | 104 | |
XM_005158491.4 | XP_005158548.1 | 3 - 4 | 506 | 104 | ||
XM_005158492.4 | XP_005158549.1 | 3 - 4 | 378 | 104 | ||
XM_017351598.2 | XP_017207087.1 | 3 - 4 | 750 | 104 | ||
GenBank mRNA | BC046874.1 | - | 3 - 4 | 231 | 104 | |
BC165673.1 | - | 2 - 3 | 148 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |