Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nup35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200066.1 | NP_956360.1 | 3 - 4 | 346 | 76 | |
XM_009304508.3 | XP_009302783.1 | 3 - 4 | 455 | 76 | ||
GenBank mRNA | BC061958.1 | - | 3 - 4 | 346 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |