Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tia1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200182.2 | NP_956476.1 | 3 - 4 | 347 | 68 | |
XM_005165110.4 | XP_005165167.1 | 3 - 4 | 404 | 68 | ||
GenBank mRNA | AY398424.1 | - | 3 - 4 | 414 | 68 | |
BC045485.1 | - | 3 - 4 | 386 | 68 | ||
BC066734.1 | - | 3 - 4 | 347 | 68 | ||
BC155269.1 | - | 3 - 4 | 350 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |