Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200241.2 | NP_956535.2 | 1 - 2 | 250 | 107 | |
GenBank mRNA | BC047810.1 | - | 1 - 2 | 382 | 107 | |
BC124685.1 | - | 1 - 2 | 246 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |