Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: strap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200304.1 | NP_956598.1 | 9 - 10 | 1131 | 111 | |
GenBank mRNA | BC049525.1 | - | 9 - 10 | 1131 | 111 | |
BC065428.1 | - | 9 - 10 | 1001 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |