Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200631.1 | NP_956925.1 | 701 | 82 | ||
GenBank mRNA | BC057414.1 | - | 701 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio |