Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bysl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201106.1 | NP_957400.1 | 2 - 3 | 446 | 80 | |
GenBank mRNA | AY648821.1 | - | 2 - 3 | 468 | 80 | |
BC053134.1 | - | 2 - 3 | 446 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |