Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scamp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201194.2 | NP_957488.1 | 2 - 3 | 206 | 77 | |
GenBank mRNA | BC055274.1 | - | 2 - 3 | 206 | 77 | |
BC056762.1 | - | 2 - 3 | 204 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |