Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arpc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201343.2 | NP_958500.1 | 3 - 4 | 367 | 98 | |
GenBank mRNA | AY398313.1 | - | 3 - 4 | 365 | 98 | |
BC048036.1 | - | 3 - 4 | 358 | 98 | ||
BC164199.1 | - | 2 - 3 | 204 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |