Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201458.2 | NP_958866.1 | 3 - 4 | 250 | 62 | |
GenBank mRNA | BC046072.1 | - | 3 - 4 | 251 | 62 | |
BC063234.1 | - | 3 - 4 | 211 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |