Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205588.3 | NP_991151.3 | 4 - 5 | 599 | 73 | |
XM_021479142.1 | XP_021334817.1 | 5 - 6 | 679 | 73 | ||
GenBank mRNA | AF510720.1 | - | 4 - 5 | 600 | 73 | |
BC094967.1 | - | 4 - 5 | 613 | 73 | ||
BC164351.1 | - | 4 - 5 | 623 | 73 | ||
EH547457.1 | - | 4 - 5 | 637 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |