Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cav3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205738.1 | NP_991301.1 | 1 - 2 | 355 | 59 | |
GenBank mRNA | AY124574.1 | - | 1 - 2 | 355 | 59 | |
BC094959.1 | - | 1 - 2 | 180 | 59 | ||
BC164895.1 | - | 1 - 2 | 143 | 59 | ||
DQ015873.1 | - | 1 - 2 | 216 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |