Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205766.1 | NP_991329.1 | 8 - 9 | 1519 | 67 | |
GenBank mRNA | BC044545.1 | - | 8 - 9 | 1519 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |