Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptk2bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212570.1 | NP_997735.1 | 5 - 6 | 705 | 78 | |
GenBank mRNA | AJ490510.2 | - | 5 - 6 | 705 | 78 | |
BC134027.1 | - | 6 - 7 | 736 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |