Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anp32b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212603.2 | NP_997768.2 | 3 - 4 | 486 | 103 | |
XM_005159865.4 | XP_005159922.1 | 3 - 4 | 1714 | 103 | ||
GenBank mRNA | BC065856.1 | - | 3 - 4 | 432 | 103 | |
CV118158.1 | - | 3 - 4 | 437 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |