Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tgm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212656.2 | NP_997821.2 | 2 - 3 | 219 | 115 | |
GenBank mRNA | AF242545.2 | - | 2 - 3 | 242 | 115 | |
BC054923.1 | - | 2 - 3 | 250 | 115 | ||
BC117635.1 | - | 2 - 3 | 219 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |