Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212682.1 | NP_997847.1 | 5 - 6 | 847 | 65 | |
GenBank mRNA | BC066618.1 | - | 5 - 6 | 847 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |