Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ythdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212713.1 | NP_997878.1 | 2 - 3 | 383 | 82 | |
XM_005167004.4 | XP_005167061.1 | 2 - 3 | 452 | 82 | ||
GenBank mRNA | BC046885.1 | - | 2 - 3 | 383 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |