Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nf2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212951.1 | NP_998116.1 | 8 - 9 | 894 | 82 | |
GenBank mRNA | AY561520.1 | - | 8 - 9 | 894 | 82 | |
BC079503.1 | - | 8 - 9 | 831 | 82 | ||
BC108005.1 | - | 8 - 9 | 900 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |