Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nf2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212951.1 | NP_998116.1 | 12 - 13 | 1354 | 57 | |
GenBank mRNA | AY561520.1 | - | 12 - 13 | 1354 | 57 | |
BC108005.1 | - | 12 - 13 | 1360 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |