Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pak5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212962.1 | NP_998127.1 | 5 - 6 | 1984 | 93 | |
GenBank mRNA | BC068016.1 | - | 5 - 6 | 1984 | 93 | |
BC168506.1 | - | 5 - 6 | 1879 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |