Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a25a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213257.1 | NP_998422.1 | 1 - 2 | 223 | 78 | |
GenBank mRNA | BC066404.1 | - | 1 - 2 | 223 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |