Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213299.1 | NP_998464.1 | 2 - 3 | 231 | 118 | |
XM_005165871.3 | XP_005165928.1 | 3 - 4 | 775 | 118 | ||
XM_005165873.3 | XP_005165930.1 | 3 - 4 | 771 | 118 | ||
GenBank mRNA | BC068387.1 | - | 2 - 3 | 231 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |