Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stx12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002051.1 | NP_001002051.1 | 5 - 6 | 528 | 89 | |
XM_009294127.3 | XP_009292402.1 | 5 - 6 | 583 | 89 | ||
XM_009294128.2 | XP_009292403.1 | 5 - 6 | 531 | 89 | ||
GenBank mRNA | BC071327.1 | - | 5 - 6 | 528 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |