Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acp5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002452.1 | NP_001002452.1 | 1 - 2 | 209 | 63 | |
GenBank mRNA | BC076019.1 | - | 1 - 2 | 209 | 63 | |
HM239641.1 | - | 1 - 2 | 41 | 63 | ||
HM239642.1 | - | 1 - 2 | 40 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |