Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab11fip4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002533.2 | NP_001002533.2 | 5 - 6 | 809 | 70 | |
XM_009299531.3 | XP_009297806.1 | 5 - 6 | 807 | 70 | ||
XM_009299532.3 | XP_009297807.1 | 3 - 4 | 717 | 70 | ||
GenBank mRNA | AB208638.1 | - | 5 - 6 | 809 | 70 | |
AB208639.1 | - | 3 - 4 | 681 | 70 | ||
BC076363.1 | - | 5 - 6 | 791 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |